Tisk…

Sulfid molybdeničitý ≥98% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Molybdenum disulphide

41827.14 41827.22 41827.36
41827.14EA 1200 CZK
41827.14 41827.22 41827.36
Sulfid molybdeničitý ≥98% (metals basis)
Sulfid molybdeničitý

Molybdenum(IV) sulfide is used as a catalyst in the electrolysis of water. It is employed as a cocatalyst for desulfurization in petrochemistry especially hydrodesulfurization. It is also used as a component of photoelectrochemical and microelectronics applications. Further, it is useful as a filament material in many electrical applications. It finds application as a lubricant in aircraft engines. Polymers consisting of molybdenum(IV) sulfide such as nylon, teflon and vespel posses improved strength and reduced friction.


  • -325 Mesh Powder


Purity: 99% CAS No.: 1317-33-5 Molecular Formula: MoS2 Formula Weight: 160.08 Physical Form: -325 Mesh Powder Appearance: Black Odour: Odourless Melting Point: 1185° Merck Reference: 14,6236 MDL No.: MFCD00003470

Vzorec: MoS₂
Bod tání: 1185 °C
Hustota: 4,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003470
CAS číslo: 1317-33-5
EINECS: 215-263-9
Merck Index: 14,6236

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR