Tisk…

Oxid wolframový ≥99.8% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Tungsten (VI) oxide

11828.22 11828.36 11828.A3
11828.22EA 2650 CZK
11828.22 11828.36 11828.A3
Oxid wolframový ≥99.8% (metals basis)
Oxid wolframový

Purity: 99.8% CAS No.: 1314-35-8 Molecular Formula: O3W Formula Weight: 231.85 Physical Form: 10-20 Micron Powder Density: 7.16 Appearance: Yellow Odour: Odourless Melting Point: 1473° Solubility: Insoluble in water and acids. Soluble in hot alkalies. Slightly soluble in HF Merck Reference: 14,9817 MDL No.: MFCD00011466


In manufacture of tungsten and tungstates, and in alloys, in fire-proofing fabrics, and as a ceramic pigment

Vzorec: O₃W
MW: 231.84
Bod tání: 1473
Hustota: 7.16
Storage Temperature: Ambient
MDL Number: MFCD00011466
CAS číslo: 1314-35-8
EINECS: 215-231-4
Merck Index: 14,9817

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR