Tisk…

Vanadium(IV) oxide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Vanadium dioxide

M-Clarity™ quality level = MQ100

215821-20G 215821-5G
SIAL215821-20GEA 11500 CZK
SIAL215821-20G SIAL215821-5G
Vanadium(IV) oxide, Sigma-Aldrich®
Vanadium(IV) oxide
Vzorec: O₂V
MW: 82,94 g/mol
Bod varu: 2700 °C (1013 hPa)
Bod tání: 238 °C
Hustota: 4,33 g/cm³ (20 °C)
MDL Number: MFCD00049702
CAS číslo: 12036-21-4
EINECS: 234-841-1
UN: 3285
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR