Tisk…

Vanadium(IV) oxide ≥99% (metals basis)

Dodavatel: Thermo Scientific
Varování

Synonyms: Vanadium dioxide

22957.09 22957.18
22957.09EA 3230 CZK
22957.09 22957.18
Vanadium(IV) oxide ≥99% (metals basis)
Vanadium(IV) oxide

Purity: 99% CAS No.: 12036-21-4 Molecular Formula: O2V, may contain V6O13 Formula Weight: 82.94 Physical Form: -100 Mesh Powder Density: 4.339 Appearance: Black Melting Point: 1967° UN No.: 3285 MDL No.: MFCD00049702

Vzorec: O₂V
MW: 82,94 g/mol
Bod varu: 2700 °C (1013 hPa)
Bod tání: 1967 °C
Hustota: 4,339
Storage Temperature: Ambient
MDL Number: MFCD00049702
CAS číslo: 12036-21-4
EINECS: 234-841-1
UN: 3285
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR