Vanadium(III) oxide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
215988-100G
215988-25G
SIAL215988-100GEA
15340
CZK
SIAL215988-100G
SIAL215988-25G
Vanadium(III) oxide, Sigma-Aldrich®
Vanadium(III) oxide
Vzorec:
O₃V₂ MW: 149,88 g/mol Bod tání: 1940 °C Hustota: 4,87 g/cm³ (20 °C) |
MDL Number:
MFCD00011455 CAS číslo: 1314-34-7 EINECS: 215-230-9 UN: 3285 ADR: 6.1,II Merck Index: 12,10059 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...