Tisk…

Vanadium(III) oxide ≥99.7% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

43377.22 43377.14
43377.22EA 12290 CZK
43377.22 43377.14
Vanadium(III) oxide ≥99.7% (metals basis)
Vanadium(III) oxide

Purity: 99.7% CAS No.: 1314-34-7 Molecular Formula: V2O3 Formula Weight: 149.88 Physical Form: Powder Density: 4.87 Appearance: Black Odour: Odourless Melting Point: 1970° Merck Reference: 14,9923 UN No.: 3285 MDL No.: MFCD00011455

Vzorec: O₃V₂
Bod tání: 1970 °C
Hustota: 4,87
Storage Temperature: Ambient
MDL Number: MFCD00011455
CAS číslo: 1314-34-7
EINECS: 215-230-9
UN: 3285
ADR: 6.1,III
Merck Index: 14,9923

Objednat

Výsledky specifikačního testu

Total Metal Impurities 0,3% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR