Tisk…

Tantalum pentafluoride ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

14052.09 14052.18
14052.09EA 6420 CZK
14052.09 14052.18
Tantalum pentafluoride ≥99.9% (metals basis)
Tantalum pentafluoride

Purity: 99.9% CAS No.: 7783-71-3 Molecular Formula: TaF5 Formula Weight: 275.94 Physical Form: Powder Density: 4.74 Appearance: White Melting Point: 96.8° Boiling Point: 229.5° Solubility: Decomposes in water forming tantalum oxyfluoride. Soluble in conc. and fuming HNO3 Merck Reference: 14,9055 UN No.: 3260 MDL No.: MFCD00042542

Vzorec: F₅Ta
Bod varu: 229,5 °C
Hustota: 4,74
Storage Temperature: Ambient
MDL Number: MFCD00042542
CAS číslo: 7783-71-3
EINECS: 232-022-3
UN: 3260
ADR: 8,II
Merck Index: 14,9055

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR