Niobium(V) fluoride 99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
87221.09
87221.18
87221.09EA
4910
CZK
87221.09
87221.18
Niobium(V) fluoride 99%
Niobium(V) fluoride
Purity: 99%
CAS No.: 7783-68-8
Molecular Formula: NbF5
Formula Weight: 187.91
Physical Form: Powder
Density: 3.92
Appearance: White to pale yellow
Melting Point: 80°
Boiling Point: ca 230°
Solubility: Hydrolyzes in water, alcohol, and caustic solutions. Sparingly soluble in CS2, chloroform. Forms complexes with Lewis bases
Merck Reference: 14,6558
UN No.: 3260
MDL No.: MFCD00011129
Vzorec:
F₅Nb MW: 187,9 g/mol Bod varu: ∼230 °C Bod tání: 80 °C Hustota: 3,92 Storage Temperature: Ambient |
MDL Number:
MFCD00011129 CAS číslo: 7783-68-8 EINECS: 232-020-2 UN: 3260 ADR: 8,II Merck Index: 14,6558 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...