Tisk…

Vanadium(III) chloride ≥99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

14039.09 14039.18
14039.09EA 2870 CZK
14039.09 14039.18
Vanadium(III) chloride ≥99% (metals basis)
Vanadium(III) chloride

Purity: 99% CAS No.: 7718-98-1 Molecular Formula: VCl3 Formula Weight: 157.30 Physical Form: -4 Mesh Density: 3.00 Appearance: Pink to purple Odour: Acrid UN No.: 2475 MDL No.: MFCD00011454

Vzorec: Cl₃V
Hustota: 3,00
Storage Temperature: Ambient
MDL Number: MFCD00011454
CAS číslo: 7718-98-1
EINECS: 231-744-6
UN: 2475
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR