Tisk…

Tantalum pentachloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Tantalum (V) chloride

M-Clarity™ quality level = MQ100

510688-25G 510688-5G
SIAL510688-25GEA 11050 CZK
SIAL510688-25G SIAL510688-5G
Tantalum pentachloride, Sigma-Aldrich®
Tantalum pentachloride
Vzorec: Cl₅Ta
MW: 358,21 g/mol
Bod varu: 232…235 °C (1013 hPa)
Bod tání: 216…217 °C
Hustota: 3,68 g/cm³ (20 °C)
MDL Number: MFCD00011253
CAS číslo: 7721-01-9
EINECS: 231-755-6
UN: 3260
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR