Tisk…

Tantalum pentachloride ≥99.99% (metals basis), Puratronic®

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Tantalum (V) chloride

Ampuled under argon

14708.14 14708.22 14708.06
14708.14EA 6180 CZK
14708.14 14708.22 14708.06
Tantalum pentachloride ≥99.99% (metals basis), Puratronic®
Tantalum pentachloride

Purity: 99.99% CAS No.: 7721-01-9 Molecular Formula: TaCl5 Formula Weight: 358.21 Physical Form: -60 Mesh Resublimed Powder, Ampouled under argon Density: 3.68 Appearance: White to pale yellow Odour: Acidic Melting Point: 216° Boiling Point: 242° Merck Reference: 14,9054 UN No.: 3260 MDL No.: MFCD00011253

Vzorec: Cl₅Ta
MW: 358,21 g/mol
Bod varu: 242 °C
Bod tání: 216 °C
Hustota: 3,68
Storage Temperature: Ambient
MDL Number: MFCD00011253
CAS číslo: 7721-01-9
EINECS: 231-755-6
UN: 3260
ADR: 8,III
Merck Index: 14,9054

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR