Tisk…

Niobium(V) chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Niobium pentachloride

M-Clarity™ quality level = MQ100

510696-25G 510696-5G
SIAL510696-25GEA 13870 CZK
SIAL510696-25G SIAL510696-5G
Niobium(V) chloride, Sigma-Aldrich®
Niobium(V) chloride
Vzorec: Cl₅Nb
MW: 270.17
Bod varu: 254
Bod tání: 205
Hustota: 2.75 g/cm³ (20 °C)
MDL Number: MFCD00011127
CAS číslo: 10026-12-7
EINECS: 233-059-8
UN: 3260
ADR: 8 III
Merck Index: 12,06649

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR