Niobium(V) chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Niobium pentachloride
M-Clarity™ quality level = MQ100
336602-10G
336602-50G
SIAL336602-10GEA
3670
CZK
SIAL336602-10G
SIAL336602-50G
Niobium(V) chloride, Sigma-Aldrich®
Niobium(V) chloride
Vzorec:
Cl₅Nb Bod varu: 254 °C (1013 hPa) Bod tání: 205 °C Hustota: 2,75 g/cm³ (20 °C) |
MDL Number:
MFCD00011127 CAS číslo: 10026-12-7 EINECS: 233-059-8 UN: 3260 ADR: 8,III Merck Index: 12,06649 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...