Niobium(V) chloride ≥99.999% (metals basis), Puratronic®
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Niobium pentachloride
Packaged under argon.
43729.22
43729.14
43729.06
43729.22EA
41110
CZK
43729.22
43729.14
43729.06
Niobium(V) chloride ≥99.999% (metals basis), Puratronic®
Niobium(V) chloride
Purity: 99.999%
CAS No.: 10026-12-7
Molecular Formula: NbCl5
Formula Weight: 270.20
Physical Form: Powder, Packaged under argon
Density: 2.74
Melting Point: 208.3°
Boiling Point: 248.2°
Merck Reference: 14,6557
UN No.: 3260
MDL No.: MFCD00011127
Vzorec:
Cl₅Nb MW: 270.17 Bod varu: 248.2 °C Bod tání: 208.3 °C Hustota: 2.74 Storage Temperature: Ambient |
MDL Number:
MFCD00011127 CAS číslo: 10026-12-7 EINECS: 233-059-8 UN: 3260 ADR: 8 III Merck Index: 14,6557 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...