Tisk…

Niobium(V) chloride ≥99.95% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Packaged under argon.

35795.09 35795.18
35795.09EA 2200 CZK
35795.09 35795.18
Niobium(V) chloride ≥99.95% (metals basis)
Niobium(V) chloride

Purity: 99.95% CAS No.: 10026-12-7 Molecular Formula: NbCl5 Formula Weight: 270.20 Physical Form: Crystalline, Packaged under argon Density: 2.74 Melting Point: 208.3° Boiling Point: 248.2° Merck Reference: 14,6557 UN No.: 3260 MDL No.: MFCD00011127

Vzorec: Cl₅Nb
Bod varu: 248,2 °C
Hustota: 2,74
Storage Temperature: Ambient
MDL Number: MFCD00011127
CAS číslo: 10026-12-7
EINECS: 233-059-8
UN: 3260
ADR: 8,III
Merck Index: 14,6557

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR