Titanium(IV) sulphide ≥99.8% (metals basis)
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Titanium disulphide
12826.06
12826.14
12826.06EA
9410
CZK
12826.06
12826.14
Titanium(IV) sulphide ≥99.8% (metals basis)
Titanium(IV) sulphide
Purity: 99.8%
CAS No.: 12039-13-3
Molecular Formula: TiS2 (approx.)
Formula Weight: 112.03
Physical Form: -200 Mesh Powder
Sensitivity: Moisture Sensitive
UN No.: 3134
MDL No.: MFCD00049598
Vzorec:
S₂Ti MW: 112,01 g/mol Hustota: 3,22 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049598 CAS číslo: 12039-13-3 EINECS: 234-883-0 UN: 3174 ADR: 4.2,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...