Tisk…

Titanium(IV) methoxide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Titanium tetramethanolate, Titanium (IV) methoxide

M-Clarity™ quality level = MQ100

404950-10G 404950-50G
SIAL404950-10GEA 2340 CZK
SIAL404950-10G SIAL404950-50G
Titanium(IV) methoxide, Sigma-Aldrich®
Titanium(IV) methoxide
Vzorec: C₄H₁₂O₄Ti
MW: 172,02 g/mol
MDL Number: MFCD00014883
CAS číslo: 992-92-7
EINECS: 213-596-4
UN: 3181
ADR: 4.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR