Titanium(IV) oxide anatase, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Pigment White 6, Titanium dioxide anatase, Anatase
M-Clarity™ quality level = MQ100
637254-100G
637254-500G
637254-50G
SIAL637254-100GEA
10590
CZK
SIAL637254-100G
SIAL637254-500G
SIAL637254-50G
Titanium(IV) oxide anatase, Sigma-Aldrich®
Titanium(IV) oxide anatase
Vzorec:
TiO₂ MW: 79.88 |
MDL Number:
MFCD00011269 CAS číslo: 1317-70-0 EINECS: 215-280-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...