Titanium(IV) oxide anatase, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Pigment White 6, Titanium dioxide anatase, Anatase
M-Clarity™ quality level = MQ200
248576-100G
248576-10KG
248576-1KG
SIAL248576-100GEA
1110
CZK
SIAL248576-100G
SIAL248576-10KG
SIAL248576-1KG
Titanium(IV) oxide anatase, Sigma-Aldrich®
Titanium(IV) oxide anatase
Vzorec:
TiO₂ MW: 79,88 g/mol |
MDL Number:
MFCD00011269 CAS číslo: 1317-70-0 EINECS: 215-280-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...