Tisk…

Titanium(IV) oxide anatase ≥99.6% (metals basis)

Dodavatel: Thermo Scientific
Varování

Synonyms: Pigment White 6, Titanium dioxide anatase, Anatase

36199.14 36199.22 36199.36
36199.14EA 1530 CZK
36199.14 36199.22 36199.36
Titanium(IV) oxide anatase ≥99.6% (metals basis)
Titanium(IV) oxide anatase

Purity: 99.6% CAS No.: 1317-70-0 Molecular Formula: O2Ti Formula Weight: 79.90 Physical Form: -325 Mesh Powder Density: 3.9 Merck Reference: 14,9472 MDL No.: MFCD00011269

Vzorec: TiO₂
MW: 79,88 g/mol
Hustota: 3,9
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS číslo: 1317-70-0
EINECS: 215-280-1
Merck Index: 14,9472

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR