Oxid titanatý ≥99.9% (metals basis)
Dodavatel: Thermo Fisher Scientific
Synonyms:
Titanium oxide, Titanium monoxide
42939.14
42939.22
42939.36
42939.14EA
3780
CZK
42939.14
42939.22
42939.36
Oxid titanatý ≥99.9% (metals basis)
Oxid titanatý
Purity: 99.9%
CAS No.: 12137-20-1
Molecular Formula: TiO
Formula Weight: 63.90
Physical Form: Lump
Density: 4.95
Melting Point: 1700°
Boiling Point:
Vzorec:
OTi MW: 63.88 Bod varu: >3000 °C Bod tání: 1700 °C Hustota: 4.95 Storage Temperature: Ambient |
MDL Number:
MFCD00036279 CAS číslo: 12137-20-1 EINECS: 235-236-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...