Hafnium dioxide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
202118-100G
202118-25G
SIAL202118-100GEA
9120
CZK
SIAL202118-100G
SIAL202118-25G
Hafnium dioxide, Sigma-Aldrich®
Hafnium dioxide
Vzorec:
HfO₂ MW: 210.49 Bod varu: 5400 Bod tání: 2812 Hustota: 9.68 g/cm³ (20 °C) |
MDL Number:
MFCD00003565 CAS číslo: 12055-23-1 EINECS: 235-013-2 Merck Index: 14,04727 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...