Tisk…

Hafnium dioxide, tech.

Dodavatel: Thermo Fisher Scientific

40270.22 40270.36
40270.22EA 6760 CZK
40270.22 40270.36
Hafnium dioxide, tech.
Hafnium dioxide

CAS No.: 12055-23-1 Molecular Formula: HfO2 Formula Weight: 210.49 Physical Form: -325 Mesh Powder Density: 9.68 Odour: Odourless Merck Reference: 14,4588 MDL No.: MFCD00003565

Vzorec: HfO₂
MW: 210.49
Bod varu: 5400
Bod tání: 2774 °C
Hustota: 9.68
Storage Temperature: Ambient
MDL Number: MFCD00003565
CAS číslo: 12055-23-1
EINECS: 235-013-2
Merck Index: 14,4588

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR