Hydrid titanatý ≥99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Titanium hydride, Titanium dihydride
12857.06
12857.22
12857.36
12857.D3
12857.06EA
1690
CZK
12857.06
12857.22
12857.36
12857.D3
Hydrid titanatý ≥99% (metals basis)
Hydrid titanatý
Purity: 99%
CAS No.: 7704-98-5
Molecular Formula: H2Ti
Formula Weight: 49.92
Physical Form: -325 Mesh Powder
Density: 3.9
Appearance: Dark grey
Odour: Odourless
Melting Point: 450° dec.
Merck Reference: 14,9473
UN No.: 1871
MDL No.: MFCD00151301
Vzorec:
H₂Ti MW: 49.9 Bod tání: 450 °C (rozkl.) Hustota: 3.9 Storage Temperature: Ambient |
MDL Number:
MFCD00151301 CAS číslo: 7704-98-5 EINECS: 231-726-8 UN: 1871 ADR: 4.1 II Merck Index: 14,9473 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...