Zirconium tetrafluoride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Zirconium (IV) fluoride
M-Clarity™ quality level = MQ100
311464-100G
311464-25G
SIAL311464-100GEA
8760
CZK
SIAL311464-100G
SIAL311464-25G
Zirconium tetrafluoride, Sigma-Aldrich®
Zirconium tetrafluoride
Vzorec:
F₄Zr MW: 167.22 Hustota: 4.43 g/cm³ (20 °C) |
MDL Number:
MFCD00011307 CAS číslo: 7783-64-4 EINECS: 232-018-1 UN: 3260 ADR: 8 II Merck Index: 12,10308 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...