Tisk…

Titanium(IV) fluoride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

333239-100G 333239-10G
SIAL333239-100GEA 11200 CZK
SIAL333239-100G SIAL333239-10G
Titanium(IV) fluoride, Sigma-Aldrich®
Titanium(IV) fluoride
Vzorec: F₄Ti
MW: 123,87 g/mol
Bod varu: 284 °C (1013 hPa)
Bod tání: 400 °C
Hustota: 2,798 g/cm³ (20.5 °C)
MDL Number: MFCD00042545
CAS číslo: 7783-63-3
EINECS: 232-017-6
UN: 3260
ADR: 8,II
Merck Index: 12,09619

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR