Hafnium(IV) chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Hafnium tetrachloride
M-Clarity™ quality level = MQ100
258202-10G
258202-50G
SIAL258202-10GEA
1260
CZK
SIAL258202-10G
SIAL258202-50G
Hafnium(IV) chloride, Sigma-Aldrich®
Hafnium(IV) chloride
Vzorec:
Cl₄Hf |
MDL Number:
MFCD00011034 CAS číslo: 13499-05-3 EINECS: 236-826-5 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...