Tisk…

Titanium(IV) bromide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Titanium tetrabromide

M-Clarity™ quality level = MQ100

307793-100G 307793-25G
SIAL307793-100GEA 8570 CZK
SIAL307793-100G SIAL307793-25G
Titanium(IV) bromide, Sigma-Aldrich®
Titanium(IV) bromide
Vzorec: Br₄Ti
Bod varu: 230 °C (1013 hPa)
Bod tání: 39 °C
Hustota: 2,6 g/cm³ (20 °C)
MDL Number: MFCD00011265
CAS číslo: 7789-68-6
EINECS: 232-185-0
UN: 3260
ADR: 8,II
Merck Index: 12,09617

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR