Scandium(III) iodide, ultra dry ≥99.999% (REO, rare earth oxide basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Ampuled under argon
13687.03
13687.06
13687.03EA
10850
CZK
13687.03
13687.06
Scandium(III) iodide, ultra dry ≥99.999% (REO, rare earth oxide basis)
Scandium(III) iodide
Purity: 99.999%
CAS No.: 14474-33-0
Molecular Formula: ScI3
Formula Weight: 425.67
Physical Form: Powder, Ampouled under argon
Sensitivity: Hygroscopic
MDL No.: MFCD00054037
Vzorec:
I₃Sc Bod tání: 945 °C Storage Temperature: Ambient |
MDL Number:
MFCD00054037 CAS číslo: 14474-33-0 EINECS: 238-469-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...