Yttrium(III) chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
451363-10G
451363-50G
SIAL451363-10GEA
3830
CZK
SIAL451363-10G
SIAL451363-50G
Yttrium(III) chloride, Sigma-Aldrich®
Yttrium(III) chloride
Vzorec:
Cl₃Y MW: 195.26 Bod varu: 1488 Bod tání: 703 Hustota: 2.52 g/cm³ (700 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011470 CAS číslo: 10361-92-9 EINECS: 233-801-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...