Yttrium(III) chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
M-Clarity™ quality level = MQ100
450103-25G
450103-5G
SIAL450103-25GEA
13140
CZK
SIAL450103-25G
SIAL450103-5G
Yttrium(III) chloride, Sigma-Aldrich®
Yttrium(III) chloride
Vzorec:
Cl₃Y MW: 195,26 g/mol Bod varu: 1488 °C (1013 hPa) Bod tání: 703 °C Hustota: 2,52 g/cm³ (700 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011470 CAS číslo: 10361-92-9 EINECS: 233-801-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...