Tisk…

Yttrium(III) bromide, ultra dry ≥99.9% (REO, rare earth oxide basis)

Dodavatel: Thermo Scientific
Varování


Ampuled under argon

35746.04 35746.09
35746.04EA 4950 CZK
35746.04 35746.09
Yttrium(III) bromide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Yttrium(III) bromide

Purity: 99.9% CAS No.: 13469-98-2 Molecular Formula: YBr3 Formula Weight: 328.63 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00049710

Vzorec: Br₃Y
MW: 328,62 g/mol
Bod tání: 904 °C
Storage Temperature: Ambient
MDL Number: MFCD00049710
CAS číslo: 13469-98-2
EINECS: 236-728-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR