Yttrium(III) bromide, hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Dodavatel: Thermo Fisher Scientific
|
Varování
|
13158.14
13158.22
13158.14EA
4840
CZK
13158.14
13158.22
Yttrium(III) bromide, hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Yttrium(III) bromide
Purity: 99.99%
Molecular Formula: YBr3.xH2O
Formula Weight: 328.63(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Upozornení: Hygroscopic. Store in cool, dry, conditions in well sealed container. Incompatible with water, moisture.
Vzorec:
YBr₃·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00049710 CAS číslo: 13469-98-2 (anhy) EINECS: 236-728-2 |
Výsledky specifikačního testu
Total Rare Earth Oxide impurities | 0.01% max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...