Cadmium(II) telluride, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Cadmium telluride
M-Clarity™ quality level = MQ100
256544-10G
256544-50G
SIAL256544-10GEA
3660
CZK
SIAL256544-10G
SIAL256544-50G
Cadmium(II) telluride, Sigma-Aldrich®
Cadmium(II) telluride
Vzorec:
CdTe MW: 240,01 g/mol Bod tání: 1089,85 °C Hustota: 6,2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00015998 CAS číslo: 1306-25-8 EINECS: 215-149-9 UN: 2570 ADR: 6.1,III Merck Index: 12,01667 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...