Síran rtuťnatý ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Mercury sulphate, Mercuric sulphate
A16330.0B
A16330.18
A16330.30
A16330.0BEA
19940
CZK
A16330.0B
A16330.18
A16330.30
Síran rtuťnatý ≥98%
Síran rtuťnatý
Purity: 98+%
CAS No.: 7783-35-9
Molecular Formula: HgO4S
Formula Weight: 296.65
Melting Point: dec.
Merck Reference: 14,5887
MDL No.: MFCD00011047
Vzorec:
HgSO₄ Bod tání: Dec. Hustota: 6,470 Storage Temperature: Ambient |
MDL Number:
MFCD00011047 CAS číslo: 7783-35-9 EINECS: 231-992-5 UN: 1645 ADR: 6.1,II Merck Index: 14,5887 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...