Tisk…

Cadmium(II) sulphate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

481882-25G 481882-5G
SIAL481882-25GEA 8430 CZK
SIAL481882-25G SIAL481882-5G
Cadmium(II) sulphate, Sigma-Aldrich®
Cadmium(II) sulphate
Vzorec: CdO₄S
Bod tání: 1000 °C
Hustota: 4,7 g/cm³ (20 °C)
MDL Number: MFCD00010923
CAS číslo: 10124-36-4
EINECS: 233-331-6
UN: 2570
ADR: 6.1,III
Merck Index: 12,01665

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR