Zinc selenide ≥99.99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
41958.14
41958.22
41958.14EA
6110
CZK
41958.14
41958.22
Zinc selenide ≥99.99% (metals basis)
Zinc selenide
Purity: 99.99%
CAS No.: 1315-09-9
Molecular Formula: ZnSe
Formula Weight: 144.33
Physical Form: -325 Mesh Powder
Density: 5.42
Melting Point:
Vzorec:
SeZn Bod tání: >1100 °C Hustota: 5,42 Storage Temperature: Ambient |
MDL Number:
MFCD00011296 CAS číslo: 1315-09-9 EINECS: 215-259-7 UN: 3283 ADR: 6.1,III Merck Index: 14,10156 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...