Sulfid zinečnatý ≥98%
Dodavatel: Thermo Fisher Scientific
A16296.0B
A16296.0I
A16296.30
A16296.0BEA
3470
CZK
A16296.0B
A16296.0I
A16296.30
Sulfid zinečnatý ≥98%
Sulfid zinečnatý
Purity: 98%
CAS No.: 1314-98-3
Molecular Formula: SZn
Formula Weight: 97.43
Odour: Odourless
Melting Point: 1700°
MDL No.: MFCD00011301
Vzorec:
SZn Bod tání: 1700 °C Hustota: 4,10 Storage Temperature: Ambient |
MDL Number:
MFCD00011301 CAS číslo: 1314-98-3 EINECS: 215-251-3 Merck Index: 14,10160 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...