Tisk…

Sulfid zinečnatý ≥99.995% (metals basis)

Dodavatel: Thermo Fisher Scientific

45454.30 45454.18 45454.09
45454.30EA 9440 CZK
45454.30 45454.18 45454.09
Sulfid zinečnatý ≥99.995% (metals basis)
Sulfid zinečnatý

Purity: 99.995% CAS No.: 1314-98-3 Molecular Formula: SZn Formula Weight: 97.43 Physical Form: 6mm & down pieces Density: 4.10 Appearance: White Odour: Odourless Melting Point: 1700° Merck Reference: 14,10160 MDL No.: MFCD00011301

Vzorec: SZn
Bod tání: 1700 °C
Hustota: 4,10
Storage Temperature: Ambient
MDL Number: MFCD00011301
CAS číslo: 1314-98-3
EINECS: 215-251-3
Merck Index: 14,10160

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR