Tisk…

Sulfid kademnatý ≥99.99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

41511.14 41511.22
41511.14EA 13420 CZK
41511.14 41511.22
Sulfid kademnatý ≥99.99% (metals basis)
Sulfid kademnatý

Purity: 99.99% CAS No.: 1306-23-6 Molecular Formula: CdS Formula Weight: 144.46 Physical Form: 3-12mm Sintered Lump Density: 4.82 Appearance: Orange Odour: Odourless Melting Point: 980° subl. Merck Reference: 14,1628 UN No.: 3077 MDL No.: MFCD00010922


Vacuum deposition

Vzorec: CdS
Bod tání: 980 °C (subl.)
Hustota: 4,82
Storage Temperature: Ambient
MDL Number: MFCD00010922
CAS číslo: 1306-23-6
EINECS: 215-147-8
UN: 3077
ADR: 9,III
Merck Index: 14,1628

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR