Tisk…

Cadmium(II) hydroxide ≥99.98% (metals basis), Puratronic®

Dodavatel: Thermo Scientific
Varování

10662.04 10662.09 10662.14
10662.04EA 2270 CZK
10662.04 10662.09 10662.14
Cadmium(II) hydroxide ≥99.98% (metals basis), Puratronic®
Cadmium(II) hydroxide

Purity: 99.98% CAS No.: 21041-95-2 Molecular Formula: Cd(OH)2 Formula Weight: 146.41 Physical Form: Powder Density: 4.79 Melting Point: 300° dec. Merck Reference: 14,1620 UN No.: 2570 MDL No.: MFCD00015995

Vzorec: H₂CdO₂
MW: 146,42 g/mol
Bod tání: 300 °C (rozkl.)
Hustota: 4,79
Storage Temperature: Ambient
MDL Number: MFCD00015995
CAS číslo: 21041-95-2
EINECS: 244-168-5
UN: 2570
ADR: 6.1,III
Merck Index: 14,1620

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR