Tisk…

Oxid kademnatý ≥99.95% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Cadmium (II) oxide

12219.36 12219.A3
12219.36EA 3850 CZK
12219.36 12219.A3
Oxid kademnatý ≥99.95% (metals basis)
Oxid kademnatý

Purity: 99.95% CAS No.: 1306-19-0 Molecular Formula: CdO Formula Weight: 128.40 Physical Form: -325 Mesh Powder Density: 8.15 Appearance: Red-brown Odour: Odourless Melting Point: 1430° Merck Reference: 14,1623 UN No.: 2570 MDL No.: MFCD00010921

Vzorec: CdO
MW: 128.41
Bod varu: 1559
Bod tání: 1430 °C
Hustota: 8.15
Storage Temperature: Ambient
MDL Number: MFCD00010921
CAS číslo: 1306-19-0
EINECS: 215-146-2
UN: 2570
ADR: 6.1 III
Merck Index: 14,1623

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR