Kyanid zinečnatý 98
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Zinc (II) cyanide
L15836.14
L15836.22
L15836.0B
L15836.14EA
906
CZK
L15836.14
L15836.22
L15836.0B
Kyanid zinečnatý 98
Kyanid zinečnatý
Purity: 98%
CAS No.: 557-21-1
Molecular Formula: Zn(CN)2
Formula Weight: 117.42
Melting Point: 800° dec.
Beilstein Reference: 4124366
Merck Reference: 14,10135
UN No.: 1713
MDL No.: MFCD00011292
Vzorec:
C₂N₂Zn MW: 117.43 Bod tání: 800 °C (rozkl.) Hustota: 1.852 Storage Temperature: Ambient |
MDL Number:
MFCD00011292 CAS číslo: 557-21-1 EINECS: 209-162-9 UN: 1713 Merck Index: 12,10264 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...