Bromid zinečnatý, anhydrous 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Zinc (II) bromide
B22510.22
B22510.36
B22510.22EA
1110
CZK
B22510.22
B22510.36
Bromid zinečnatý, anhydrous 98%
Bromid zinečnatý
Purity: 98%
CAS No.: 7699-45-8
Molecular Formula: ZnBr2
Formula Weight: 225.19
Refractive Index: 1.5452
Melting Point: 394°
Boiling Point: 650°
Merck Reference: 14,10129
UN No.: 3260
MDL No.: MFCD00011294
Vzorec:
Br₂Zn Bod varu: 650 °C Bod tání: 394 °C Hustota: 4,201 Storage Temperature: Ambient |
MDL Number:
MFCD00011294 CAS číslo: 7699-45-8 EINECS: 231-718-4 UN: 3260 ADR: 8,II Merck Index: 14,10129 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...