Bromid zinečnatý, ultra dry ≥99.999% (metals basis)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Zinc (II) bromide
Ampuled under argon
35725.06
35725.22
35725.06EA
3750
CZK
35725.06
35725.22
Bromid zinečnatý, ultra dry ≥99.999% (metals basis)
Bromid zinečnatý
Purity: 99.999%
CAS No.: 7699-45-8
Molecular Formula: ZnBr2
Formula Weight: 225.19
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.201
Refractive Index: 1.5452
Sensitivity: Hygroscopic
Melting Point: 394°
Boiling Point: 650°
Merck Reference: 14,10129
UN No.: 3260
MDL No.: MFCD00011294
Vzorec:
Br₂Zn MW: 225,2 g/mol Bod varu: 650 °C Bod tání: 394 °C Hustota: 4,201 Storage Temperature: Ambient |
MDL Number:
MFCD00011294 CAS číslo: 7699-45-8 EINECS: 231-718-4 UN: 3260 ADR: 8,II Merck Index: 14,10129 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...