Copper(I) thiocyanate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Copper thiocyanate, Copper (I) rhodanide, Cuprous thiocyanate, Copper rhodanide
M-Clarity™ quality level = MQ100
298212-250G
298212-50G
SIAL298212-250GEA
10990
CZK
SIAL298212-250G
SIAL298212-50G
Copper(I) thiocyanate, Sigma-Aldrich®
Copper(I) thiocyanate
Vzorec:
CCuNS MW: 121,63 g/mol Hustota: 2,846 g/cm³ (15 °C) |
MDL Number:
MFCD00010980 CAS číslo: 1111-67-7 EINECS: 214-183-1 UN: 3077 ADR: 9,III Merck Index: 13,02699 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...