Copper(I) thiocyanate ≥96%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Copper thiocyanate, Copper (I) rhodanide, Cuprous thiocyanate, Copper rhodanide
40220.18
40220.30
40220.18EA
1730
CZK
40220.18
40220.30
Copper(I) thiocyanate ≥96%
Copper(I) thiocyanate
Purity: 96% min
CAS No.: 1111-67-7
Molecular Formula: CuSCN
Formula Weight: 121.63
Physical Form: Powder
Density: 2.84
Melting Point: 1084°
Solubility: Practically insoluble in water, alcohol. Soluble in NH4OH, ether
Merck Reference: 14,2669
UN No.: 3077
MDL No.: MFCD00010980
Vzorec:
CCuNS MW: 121,63 g/mol Bod tání: 1084 °C Hustota: 2,84 Storage Temperature: Ambient |
MDL Number:
MFCD00010980 CAS číslo: 1111-67-7 EINECS: 214-183-1 UN: 3077 ADR: 9,III Merck Index: 14,2669 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...