Tisk…

Copper(II) sulphide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

342467-100G 342467-500G
SIAL342467-100GEA 4370 CZK
SIAL342467-100G SIAL342467-500G
Copper(II) sulphide, Sigma-Aldrich®
Copper (II) sulphide
Vzorec: CuS
MW: 95,61 g/mol
Hustota: 4,6 g/cm³ (20 °C)
MDL Number: MFCD00016066
CAS číslo: 1317-40-4
EINECS: 215-271-2
Merck Index: 12,02724

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR