Tisk…

Silver cyanate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

244694-10G 244694-50G
SIAL244694-10GEA 2750 CZK
SIAL244694-10G SIAL244694-50G
Silver cyanate, Sigma-Aldrich®
Silver cyanate
Vzorec: CAgNO
Hustota: 4,15 g/cm³ (20 °C)
MDL Number: MFCD00011512
CAS číslo: 3315-16-0
EINECS: 222-006-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR