Oxid měďný ≥99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Cuprous oxide, Dicopper oxide
40188.14
40188.22
40188.36
40188.A4
40188.14EA
1230
CZK
40188.14
40188.22
40188.36
40188.A4
Oxid měďný ≥99% (metals basis)
Oxid měďný
Purity: 99%
CAS No.: 1317-39-1
Molecular Formula: Cu2O
Formula Weight: 143.08
Physical Form: -200 Mesh Powder
Density: 6.0
Refractive Index: 2.705
Sensitivity: Air and Moisture Sensitive
Appearance: Red to dark red
Odour: Odourless
Melting Point: 1235°
Boiling Point: 1800° dec.
Merck Reference: 14,2664
UN No.: 3077
MDL No.: MFCD00010974
Vzorec:
Cu₂O MW: 143.09 Bod varu: 1800 °C (rozkl.) Bod tání: 1235 °C Hustota: 6.0 Storage Temperature: Ambient |
MDL Number:
MFCD00010974 CAS číslo: 1317-39-1 EINECS: 215-270-7 UN: 3077 ADR: 9 III Merck Index: 14,2664 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...